N,N-Dimethyl-1-naphthylamine 99%

Supplier: Thermo Scientific Chemicals
Warning

20417-0050 20417-0250 20417-1000 20417-5000
TS20417-0050EA 50 USD
TS20417-0050 TS20417-0250 TS20417-1000 TS20417-5000
N,N-Dimethyl-1-naphthylamine 99%
N,N-Dimethyl-1-naphthylamine
Formula: C₁₂H₁₃N
Boiling Pt: 139…140 °C (13 mmHg)
Density: 1.04 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003919
CAS Number: 86-56-6
EINECS: 201-682-4
UN: 3082
ADR: 9,III
Merck Index: 12,03301

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6210 to 1.6230 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR