Boron trifluoride dimethyl ether complex

Supplier: Thermo Scientific Chemicals
Danger

20406-0500 20406-2500
TS20406-0500EA 53 USD
TS20406-0500 TS20406-2500
Boron trifluoride dimethyl ether complex
Boron trifluoride dimethyl ether complex
Formula: C₂H₆BF₃O
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –14 °C
Density: 1.23 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
CAS Number: 353-42-4
EINECS: 206-532-1
UN: 2965
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.3070 to 1.3090 (20°C, 589 nm)
Assay ≥59 % (BF3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR