Cyclophosphamide monohydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

20396-0010 20396-0050 20396-0250
TS20396-0010EA 65 USD
TS20396-0010 TS20396-0050 TS20396-0250
Cyclophosphamide monohydrate 97%
Cyclophosphamide monohydrate

Appearance: White to off-white Crystalline powder or crystals

Formula: C₇H₁₇Cl₂N₂O₃P
Melting Pt: 49…53 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149395
CAS Number: 6055-19-2
UN: 3464
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 53 °C
Water 5.5 to 7.5 % (K.F.)
HPLC ≥96.0 % (ELSD)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR