Tetramethoxysilane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetramethyl orthosilicate
AC20382-0025
AC20382-0250
AC20382-1000
AC20382-5000
TS20382-0025EA
841
USD
TS20382-0025
TS20382-0250
TS20382-1000
TS20382-5000
Tetramethoxysilane 99%
Tetramethoxysilane
Formula:
C₄H₁₂O₄Si MW: 152.22 g/mol Boiling Pt: 121 Melting Pt: -2 Density: 1.03 g/cm³ Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008341 CAS Number: 681-84-5 EINECS: 211-656-4 UN: 2606 ADR: 6.1 I |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3670 to 1.3700 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...