Tetramethoxysilane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetramethyl orthosilicate

AC20382-0025 AC20382-0250 AC20382-1000 AC20382-5000
TS20382-0025EA 841 USD
TS20382-0025 TS20382-0250 TS20382-1000 TS20382-5000
Tetramethoxysilane 99%
Tetramethoxysilane
Formula: C₄H₁₂O₄Si
MW: 152.22 g/mol
Boiling Pt: 121
Melting Pt: -2
Density: 1.03 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00008341
CAS Number: 681-84-5
EINECS: 211-656-4
UN: 2606
ADR: 6.1 I

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3670 to 1.3700 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR