4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 98%

Supplier: Thermo Scientific Chemicals

20271-0100 20271-0500
TS20271-0100EA 158 USD
TS20271-0100 TS20271-0500
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 98%
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)
Formula: C₁₆H₁₅N₅·2HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00012681
CAS Number: 28718-90-3

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Solubility (1 % in water) Clear
HPLC ≥97.5 %
Nitrogen (N) ≥18 % (Elemental analysis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR