Potassium peroxodisulfate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC20201-0010 AC20201-0050 AC20201-0250 AC20201-2500 AC20201-5000
TS20201-0010EA 100 USD
TS20201-0010 TS20201-0050 TS20201-0250 TS20201-2500 TS20201-5000
Potassium peroxodisulfate 99+% for analysis
Potassium peroxodisulphate
Formula: K₂S₂O₈
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 13,07741

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodometric ≥99.0 %
Chloride (Cl) ≤120 ppm
Iron (Fe) ≤20 ppm
Manganese (Mn) ≤5 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR