Copper(I) iodide 99.995% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cuprous iodide
AC20150-0050
AC20150-0250
AC20150-1000
TS20150-0050EA
53
USD
TS20150-0050
TS20150-0250
TS20150-1000
Copper(I) iodide 99.995% (trace metals basis)
Copper (I) iodide
Formula:
CuI Boiling Pt: 1290 °C (1013 hPa) Melting Pt: 605 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 EINECS: 231-674-6 UN: 3077 ADR: 9,III Merck Index: 13,02692 |
Specification Test Results
Appearance | White to gray to light brown Powder |
Titration Complexometric | 31.0 to 34.0 % (Cu) |
Total trace metal impurities | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...