Sulfur 99.5+%, refined

Supplier: Thermo Scientific Chemicals
Warning

201220025 201220100 201225000
TS201220025EA 217 USD
TS201220025 TS201220100 TS201225000
Sulfur 99.5+%, refined
Sulphur
Formula: S
Boiling Pt: 445 °C (760 mmHg)
Melting Pt: 114 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059

Order Now

Specification Test Results

Appearance Yellow Powder
Assay ≥99.50 %
Water ≤0.5 %
Sulfated ash ≤0.08 %
Acidity ≤0.01 % (as H2SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR