Sulfur 99.5+%, refined
Supplier: Thermo Scientific Chemicals
|
Warning
|
201220025
201220100
201225000
TS201220025EA
217
USD
TS201220025
TS201220100
TS201225000
Sulfur 99.5+%, refined
Sulphur
Formula:
S Boiling Pt: 445 °C (760 mmHg) Melting Pt: 114 °C Flash Pt: 168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085316 CAS Number: 7704-34-9 EINECS: 231-722-6 UN: 1350 ADR: 4.1,III Merck Index: 13,09059 |
Specification Test Results
Appearance | Yellow Powder |
Assay | ≥99.50 % |
Water | ≤0.5 % |
Sulfated ash | ≤0.08 % |
Acidity | ≤0.01 % (as H2SO4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...