Orthophosphoric acid 85% (w/w) in water for analysis

Supplier: Thermo Scientific Chemicals
Danger

20114-0025 20114-5000 201140010 201140100
TS20114-0025EA 182 USD
TS20114-0025 TS20114-5000 TS201140010 TS201140100
Orthophosphoric acid 85% (w/w) in water for analysis
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 21 °C
Density: 1.68 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear viscous liquid
Color scale ≤10 APHA
Titration with NaOH ≥85 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤5 ppm
Sulfate (SO4) ≤50 ppm
PO3 and PO2 (as H3PO3) ≤0.05 %
Arsenic (As) ≤1 ppm
Cadmium (Cd) ≤10 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Manganese (Mn) ≤1 ppm
Nickel (Ni) ≤10 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR