Pyronin G, high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning

20010-0010 20010-0050 20010-0250
TS20010-0010EA 47 USD
TS20010-0010 TS20010-0050 TS20010-0250
Pyronin G, high purity biological stain
Pyronine G
Formula: C₁₇H₁₉ClN₂O
Melting Pt: 250…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0
EINECS: 202-147-8
Merck Index: 1208190

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥50 %
UV Lambda max 546 to 551 nm (in 50% ethanol)
Absorption Ratio P-15/P+15 1.23 to 1.7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR