Selenium dioxide 99.8% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

20000-0025 AC20000-0100 AC20000-1000 AC20000-5000
TS20000-0025EA 886 USD
TS20000-0025 TS20000-0100 TS20000-1000 TS20000-5000
Selenium dioxide 99.8% (trace metals basis)
Selenium dioxide
Formula: SeO₂
Melting Pt: 315 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1,II
Merck Index: 13,08508

Order Now

Specification Test Results

Appearance White to light beige Powder
Assay 70 to 71.5 % Se
Total trace metal impurities ≤0.2 %
Arsenic (As) ≤20 ppm
Cadmium (Cd) ≤5 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Mercury (Hg) ≤5 ppm
Nickel (Ni) ≤10 ppm
Tellurium (Te) ≤500 ppm
Insoluble matter ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR