Sulfur 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

19993-0500 19993-2500 199930100
TS19993-0500EA 112 USD
TS19993-0500 TS19993-2500 TS199930100
Sulfur 99.999% (trace metals basis)
Sulphur
Formula: S
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 114 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059

Order Now

Specification Test Results

Appearance Yellow Pieces
Particle size ≤12 mm
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR