Copper(I) chloride 97%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cuprous chloride, Copper chloride

AC19964-0025 AC19964-0250 AC19964-5000
TS19964-0025EA 244 USD
TS19964-0025 TS19964-0250 TS19964-5000
Copper(I) chloride 97%, pure
Copper (I) chloride
Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
Density: 4.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now

Specification Test Results

Appearance Green-gray Powder or pearls
Titration Complexometric ≥96.0 %
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤400 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR