Lanthanum(III) oxide 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
19916-0010
AC19916-2500
TS19916-0010EA
510
USD
TS19916-0010
TS19916-2500
Lanthanum(III) oxide 99.9% (trace metals basis)
Lanthanum (III) oxide
Formula:
La₂O₃ Boiling Pt: 4200 °C (760 mmHg) Melting Pt: 2315 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 |
Specification Test Results
Appearance | White to off-white Powder |
Assay | ≥99.85 % (La2O3) TREO base |
Loss on ignition | ≤3.0 % (1000°C) |
Iron(III)oxide (Fe2O3) | ≤250 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...