Zinc chloride 98.5% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride
AC19894-0025
AC19894-5000
TS19894-0025EA
557
USD
TS19894-0025
TS19894-5000
Zinc chloride 98.5% for analysis
Zinc chloride
Formula:
ZnCl₂ Boiling Pt: 732 °C (1013 hPa) Melting Pt: 293 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 13,10185 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Titration Complexometric | ≥98.0 % |
Nitrate (NO3) | ≤30 ppm |
Sulfate (SO4) | ≤20 ppm |
Ammonium (NH4) | ≤50 ppm |
Cadmium (Cd) | ≤5 ppm |
Calcium (Ca) | ≤10 ppm |
Copper (Cu) | ≤10 ppm |
Iron (Fe) | ≤5 ppm |
Lead (Pb) | ≤10 ppm |
Magnesium (Mg) | ≤100 ppm |
Potassium (K) | ≤200 ppm |
Sodium (Na) | ≤50 ppm |
Oxide chloride (ZnO) | ≤1.2 % |
Insoluble matter | ≤0.005 % |
Total nitrogen | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...