Lead 99+%, powder

Supplier: Thermo Scientific Chemicals
Danger

198110010
TS198110010EA 100 USD
TS198110010
Lead 99+%, powder
Lead
Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740 °C (760 mmHg)
Melting Pt: 327.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Grey Powder
Assay ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR