Tin(IV) chloride, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Stannic chloride, Tin tetrachloride
AC19751-0010
AC19751-0025
AC19751-0100
AC19751-1000
TS19751-0010EA
282
USD
TS19751-0010
TS19751-0025
TS19751-0100
TS19751-1000
Tin(IV) chloride, anhydrous 99%
Tin (IV) chloride
Formula:
Cl₄Sn Boiling Pt: 114 °C (760 mmHg) Melting Pt: –33 °C Density: 2.22 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011242 CAS Number: 7646-78-8 EINECS: 231-588-9 UN: 1827 ADR: 8,II Merck Index: 12,08929 |
Specification Test Results
Appearance | Colorless to light yellow Liquid (May develop some turbidity or precipitate) |
Titration Argentometric | 98.5 to 101.5 % |
Free chlorine | ≤10 ppm |
Arsenic (As) | ≤40 ppm |
Copper (Cu) | ≤5 ppm |
Iron (Fe) | ≤1 ppm |
Lead (Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...