Tin(IV) chloride, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Stannic chloride, Tin tetrachloride

AC19751-0010 AC19751-0025 AC19751-0100 AC19751-1000
TS19751-0010EA 282 USD
TS19751-0010 TS19751-0025 TS19751-0100 TS19751-1000
Tin(IV) chloride, anhydrous 99%
Tin (IV) chloride
Formula: Cl₄Sn
Boiling Pt: 114 °C (760 mmHg)
Melting Pt: –33 °C
Density: 2.22 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8,II
Merck Index: 12,08929

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid (May develop some turbidity or precipitate)
Titration Argentometric 98.5 to 101.5 %
Free chlorine ≤10 ppm
Arsenic (As) ≤40 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR