Copper(II) oxide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

AC19744-0025 AC19744-0250 AC19744-5000
TS19744-0025EA 217 USD
TS19744-0025 TS19744-0250 TS19744-5000
Copper(II) oxide 97%
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674

Order Now

Specification Test Results

Appearance Black Powder
Titration Iodometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR