Bis(triphenylphosphine)palladium(II) chloride (15% Pd) for analysis
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis(triphenylphosphine)palladium(II) dichloride
AC19732-0010
AC19732-0050
AC19732-0250
TS19732-0010EA
68
USD
TS19732-0010
TS19732-0050
TS19732-0250
Bis(triphenylphosphine)palladium(II) chloride (15% Pd) for analysis
Bis(triphenylphosphine)palladium (II) chloride
Formula:
C₃₆H₃₀Cl₂P₂Pd Melting Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009593 CAS Number: 13965-03-2 EINECS: 237-744-2 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Assay | 14.6 to 15.8 % (Pd) |
Calcium (Ca) | ≤15 ppm |
Copper (Cu) | ≤10 ppm |
Gold (Au) | ≤10 ppm |
Iridium (Ir) | ≤10 ppm |
Iron (Fe) | ≤5 ppm |
Lead (Pb) | ≤5 ppm |
Magnesium (Mg) | ≤5 ppm |
Platinum (Pt) | ≤10 ppm |
Rhodium (Rh) | ≤10 ppm |
Ruthenium (Ru) | ≤10 ppm |
Silicon (Si) | ≤10 ppm |
Silver (Ag) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...