Bis(triphenylphosphine)palladium(II) chloride (15% Pd) for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(triphenylphosphine)palladium(II) dichloride

AC19732-0010 AC19732-0050 AC19732-0250
TS19732-0010EA 68 USD
TS19732-0010 TS19732-0050 TS19732-0250
Bis(triphenylphosphine)palladium(II) chloride (15% Pd) for analysis
Bis(triphenylphosphine)palladium (II) chloride
Formula: C₃₆H₃₀Cl₂P₂Pd
Melting Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00009593
CAS Number: 13965-03-2
EINECS: 237-744-2

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Assay 14.6 to 15.8 % (Pd)
Calcium (Ca) ≤15 ppm
Copper (Cu) ≤10 ppm
Gold (Au) ≤10 ppm
Iridium (Ir) ≤10 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤5 ppm
Platinum (Pt) ≤10 ppm
Rhodium (Rh) ≤10 ppm
Ruthenium (Ru) ≤10 ppm
Silicon (Si) ≤10 ppm
Silver (Ag) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR