Molecular sieve A4 (0.4 nm, 4 Å) 8 - 12 mesh

Supplier: Thermo Scientific Chemicals

19727-0050 19727-5000 197270250
TS19727-0050EA 402 USD
TS19727-0050 TS19727-5000 TS197270250
Molecular sieve A4 (0.4 nm, 4 Å) 8 - 12 mesh
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now

Specification Test Results

Appearance Beige Beads
Bulk density 680 to 760 g/l
Residual water ≤1.5 % (550°C)
Water adsorption ≥20 % (1bar,25°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR