Sodium perchlorate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

19712-0010 19712-2500 AC19712-0051
TS19712-0010EA 359 USD
TS19712-0010 TS19712-2500 TS19712-0051
Sodium perchlorate 99+% for analysis
Sodium perchlorate
Formula: ClNaO₄
Melting Pt: 482 °C
Storage Temperature: Ambient
MDL Number: MFCD00003480
CAS Number: 7601-89-0
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 13,08726

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Assay ≥99.0 % (on dry substance)
Loss on drying ≤2.0 % (130°C, 2 h)
pH 6.0 to 8.0 (5 % in water)
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤20 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤500 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR