Sodium perchlorate 99+% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
19712-0010
19712-2500
AC19712-0051
TS19712-0010EA
359
USD
TS19712-0010
TS19712-2500
TS19712-0051
Sodium perchlorate 99+% for analysis
Sodium perchlorate
Formula:
ClNaO₄ Melting Pt: 482 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003480 CAS Number: 7601-89-0 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II Merck Index: 13,08726 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Assay | ≥99.0 % (on dry substance) |
Loss on drying | ≤2.0 % (130°C, 2 h) |
pH | 6.0 to 8.0 (5 % in water) |
Chloride (Cl) | ≤30 ppm |
Sulfate (SO4) | ≤20 ppm |
Calcium (Ca) | ≤200 ppm |
Iron (Fe) | ≤5 ppm |
Potassium (K) | ≤500 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...