Polyphosphoric acid (≥84% P₂O₅), pure

Supplier: Thermo Scientific Chemicals
Danger

196950010 196950025
TS196950010EA 86 USD
TS196950010 TS196950025
Polyphosphoric acid (≥84% P₂O₅), pure
Polyphosphoric acid
Formula: H(PO₃H)nOH
Boiling Pt: 550 °C (760 mmHg)
Density: 2.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084480
CAS Number: 8017-16-1
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear viscous liquid (May darken on storage)
Color scale ≤50 APHA
Titration with NaOH ≥82 % (as P2O5)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR