Zinc chloride 98+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

19684-0010 19684-0025 AC19684-0050 AC19684-0100
TS19684-0010EA 130 USD
TS19684-0010 TS19684-0025 TS19684-0050 TS19684-0100
Zinc chloride 98+%, extra pure
Zinc chloride
Formula: ZnCl₂
Boiling Pt: 732 °C (760 mmHg)
Melting Pt: 293 °C
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Titration Complexometric ≥98.0 %
Cadmium (Cd) ≤10 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Manganese (Mn) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR