Potassium iodate 98%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

AC19674-0025 AC19674-1000 AC19674-5000
TS19674-0025EA 740 USD
TS19674-0025 TS19674-1000 TS19674-5000
Potassium iodate 98%, extra pure
Potassium iodate
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560
Density: 3.93 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1 II
Merck Index: 13,07726

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Titration Iodometric ≥97.5 %
Loss on drying ≤0.5 % (105°C, 3 hrs)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR