Copper(II) carbonate basic Cu 54-56%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)

AC19667-0025 AC19667-0250 AC19667-5000
TS19667-0025EA 230 USD
TS19667-0025 TS19667-0250 TS19667-5000
Copper(II) carbonate basic Cu 54-56%, pure
Copper (II) carbonate basic
Formula: CuCO₃·Cu(OH)₂
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00010976
CAS Number: 12069-69-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Green Crystalline powder
Assay 54 to 56 % (Cu)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR