Bromine 99.6% analysis_for
Supplier: Thermo Scientific Chemicals
|
Danger
|
19666-2500
19666-0250
196660010
TS19666-2500EA
281
USD
TS19666-2500
TS19666-0250
TS196660010
Bromine 99.6% analysis_for
Bromine
Formula:
Br₂ MW: 159.81 g/mol Boiling Pt: 58.7 Melting Pt: -7.2 Density: 3.111 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010896 CAS Number: 7726-95-6 EINECS: 231-778-1 UN: 1744 ADR: 8 I Merck Index: 13,01378 |
Specification Test Results
Appearance | Dark red-brown Liquid |
Titration Iodometric | ≥99.5 % |
Residue after evaporation | ≤100 ppm |
Chlorine (Cl) | ≤500 ppm |
Iodine (I) | ≤10 ppm |
Heavy metals (as Pb) | ≤2 ppm |
Sulfur compounds (as SO4) | ≤30 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...