Ammonium carbonate for analysis

Supplier: Thermo Scientific Chemicals
Warning

AC19665-0010 AC19665-0025 AC19665-1000 AC19665-5000
TS19665-0010EA 103 USD
TS19665-0010 TS19665-0025 TS19665-1000 TS19665-5000
Ammonium carbonate for analysis
Ammonium carbonate
Formula: (NH₄)₂CO₃
Melting Pt: 58 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Appearance White Powder or crystals and/or chunks
Non-volatile matter ≤0.01 %
Titration with HCl ≥30.0 % (NH3)
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR