Chromium(VI) oxide 99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chromic anhydride, Chromium trioxide
AC19661-0010
AC19661-0025
AC19661-0250
AC19661-2500
TS19661-0010EA
124
USD
TS19661-0010
TS19661-0025
TS19661-0250
TS19661-2500
Chromium(VI) oxide 99.5%
Chromium (VI) oxide
Formula:
CrO₃ MW: 99.99 g/mol Melting Pt: 195 Density: 2.7 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010952 CAS Number: 1333-82-0 EINECS: 215-607-8 UN: 1463 ADR: 5.1 II REACH: 01-2119458868-17 Merck Index: 13,02256 |
Specification Test Results
Appearance | Dark red to purple Crystals or granules |
Titration Iodometric | ≥99.4 % |
Sulfate (SO4) | ≤500 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...