Potassium thiocyanate 99+% for analysis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium rhodanide
AC19658-0025
AC19658-0100
AC19658-2500
AC19658-5000
TS19658-0025EA
327
USD
TS19658-0025
TS19658-0100
TS19658-2500
TS19658-5000
Potassium thiocyanate 99+% for analysis
Potassium thiocyanate
Formula:
KSCN Melting Pt: 170…179 °C Density: 1.886 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 Merck Index: 12,07866 |
Specification Test Results
Appearance | White Crystalline powder |
Titration Argentometric | ≥99.0 % |
pH | 5.3 to 8.7 (5 % at 20°C) |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤50 ppm |
Ammonium (NH4) | ≤10 ppm |
Iron (Fe) | ≤2 ppm |
Sodium (Na) | ≤200 ppm |
Heavy metals (as Pb) | ≤2 ppm |
Iodine consuming subst. | ≤30 mg I2/100 g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...