Potassium thiocyanate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

AC19658-0025 AC19658-0100 AC19658-2500 AC19658-5000
TS19658-0025EA 327 USD
TS19658-0025 TS19658-0100 TS19658-2500 TS19658-5000
Potassium thiocyanate 99+% for analysis
Potassium thiocyanate
Formula: KSCN
Melting Pt: 170…179 °C
Density: 1.886 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥99.0 %
pH 5.3 to 8.7 (5 % at 20°C)
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Ammonium (NH4) ≤10 ppm
Iron (Fe) ≤2 ppm
Sodium (Na) ≤200 ppm
Heavy metals (as Pb) ≤2 ppm
Iodine consuming subst. ≤30 mg I2/100 g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR