Aluminium chloride, anhydrous 99%, granules, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC19578-0010 AC19578-5000
TS19578-0010EA 102 USD
TS19578-0010 TS19578-5000
Aluminium chloride, anhydrous 99%, granules, Extra Pure
Aluminium chloride
Formula: AlCl₃
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 194 °C
Density: 2.44 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II
Merck Index: 13,00335

Order Now

Specification Test Results

Appearance White to yellow to gray Granules
Titration Argentometric ≥98.5 %
Aluminium (Al) ≤150 ppm (free Al)
Chromium (Cr) ≤10 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤100 ppm
Nickel (Ni) ≤20 ppm
Zinc (Zn) ≤30 ppm
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR