Potassium hexachloroplatinate(IV) (∼40% Pt)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dipotassium hexachloroplatinate

19535-0250 AC19535-0010 AC19535-0050
TS19535-0250EA 1378 USD
TS19535-0250 TS19535-0010 TS19535-0050
Potassium hexachloroplatinate(IV) (∼40% Pt)
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 2923
ADR: 8,III
Merck Index: 13,07720

Order Now

Specification Test Results

Appearance Yellow to light orange Powder and/or chunks
Platinum (Pt) 39.4 to 40.6 % (Pt)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR