Potassium hexachloroplatinate(IV) (∼40% Pt)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dipotassium hexachloroplatinate
19535-0250
AC19535-0010
AC19535-0050
TS19535-0250EA
1378
USD
TS19535-0250
TS19535-0010
TS19535-0050
Potassium hexachloroplatinate(IV) (∼40% Pt)
Potassium hexachloroplatinate(IV)
Formula:
K₂PtCl₆ Melting Pt: 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011389 CAS Number: 16921-30-5 EINECS: 240-979-3 UN: 2923 ADR: 8,III Merck Index: 13,07720 |
Specification Test Results
Appearance | Yellow to light orange Powder and/or chunks |
Platinum (Pt) | 39.4 to 40.6 % (Pt) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...