Platinium(IV) oxide 83% Pt

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Platinum dioxide, Adam's type catalyst

AC19532-0010 AC19532-0050
TS19532-0010EA 208 USD
TS19532-0010 TS19532-0050
Platinium(IV) oxide 83% Pt
Platinium (IV) oxide
Formula: O₂Pt
MW: 227.08
Melting Pt: 450
Density: 11.8
MDL Number: MFCD00011184
CAS Number: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1 I
Merck Index: 12,07686

Order Now

Specification Test Results

Appearance Brown Powder
Assay ≥79 % (Pt)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR