Palladium(II) acetate (47,5% Pd)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Palladium diacetate, Palladium (II) acetate

AC19518-0010 AC19518-0020 AC19518-0100 AC19518-0500
TS19518-0010EA 155 USD
TS19518-0010 TS19518-0020 TS19518-0100 TS19518-0500
Palladium(II) acetate (47,5% Pd)
Palladium (II) acetate
Formula: C₄H₆O₄Pd
Melting Pt: 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now

Specification Test Results

Appearance Orange to brown Powder
Infrared spectrum Conforms
Assay 46.9 to 48.5 % (Pd)
Additional info Platinum group metals (Pt+Ru+Rh+Ir): < 300 ppm
Chloride (Cl) ≤1000 ppm
Aluminium (Al) ≤50 ppm
Calcium (Ca) ≤50 ppm
Chromium (Cr) ≤10 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Magnesium (Mg) ≤50 ppm
Manganese (Mn) ≤10 ppm
Nitrogen (N) ≤2 %
Silicon (Si) ≤50 ppm
Zinc (Zn) ≤25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR