Lithium perchlorate trihydrate 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

AC19472-0010 AC19472-0250 AC19472-2500
TS19472-0010EA 332 USD
TS19472-0010 TS19472-0250 TS19472-2500
Lithium perchlorate trihydrate 99%, extra pure
Lithium perchlorate trihydrate
Formula: ClLiO₄·3H₂O
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00149765
CAS Number: 13453-78-6
EINECS: 232-237-2
UN: 1481
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Adhering crystals and/or chunks
Titration Argentometric ≥98.5 %
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR