Thallium(I) nitrate 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC19424-0100
TS19424-0100EA 77 USD
TS19424-0100
Thallium(I) nitrate 99.99% (trace metals basis)
Thallium (I) nitrate
Formula: NO₃Tl
Boiling Pt: 433 °C (760 mmHg)
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00011280
CAS Number: 10102-45-1
UN: 2727
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White Powder or crystals
Assay 75.9 to 77.5 % (Tl)
Total trace metal impurities ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR