Ammonium hexafluorosilicate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate
AC19400-0100
AC19400-0500
AC19400-2500
TS19400-0100EA
55
USD
TS19400-0100
TS19400-0500
TS19400-2500
Ammonium hexafluorosilicate 99.999% (trace metals basis)
Ammonium hexafluorosilicate
Formula:
H₈F₆N₂Si Density: 2.01 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010887 CAS Number: 16919-19-0 UN: 2854 ADR: 6.1,III |
Specification Test Results
Appearance | White Crystalline powder and/or chunks |
Total trace metal impurities | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...