Potassium nitrate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

19380-2500 AC19380-0100 AC19380-0500
TS19380-2500EA 620 USD
TS19380-2500 TS19380-0100 TS19380-0500
Potassium nitrate 99.999% (trace metals basis)
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now

Specification Test Results

Appearance White Crystals and/or chunks
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR