Potassium nitrate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
19380-2500
AC19380-0100
AC19380-0500
TS19380-2500EA
620
USD
TS19380-2500
TS19380-0100
TS19380-0500
Potassium nitrate 99.999% (trace metals basis)
Potassium nitrate
Formula:
KNO₃ Melting Pt: 334 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 13,07733 |
Specification Test Results
Appearance | White Crystals and/or chunks |
Total trace metal impurities | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...