Platinum 99.999% (trace metals basis), powder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Platinium

193710010
TS193710010EA 369 USD
TS193710010
Platinum 99.999% (trace metals basis), powder
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827 °C (760 mmHg)
Melting Pt: 1772 °C
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR