Nickel(II) chloride hexahydrate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride hexahydrate

AC19357-0050 AC19357-0250 AC19357-1000
TS19357-0050EA 78 USD
TS19357-0050 TS19357-0250 TS19357-1000
Nickel(II) chloride hexahydrate 99.999% (trace metals basis)
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
Melting Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529

Order Now

Specification Test Results

Appearance Green Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤0.001 %
Nickel (Ni) 24 to 26 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR