Ammonium iron (II) sulfate hexahydrate 98%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate

AC19325-0050
TS19325-0050EA 155 USD
TS19325-0050
Ammonium iron (II) sulfate hexahydrate 98%, extra pure
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9

Order Now

Specification Test Results

Appearance Green to blue-green to blue to dark blue Powder
pH 3 to 5
Titration with KMnO4 ≥97.5 %
Insoluble matter ≤0.05 % (in dilute H2SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR