Bismuth(III) oxide 98+%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Bismuth Trioxide, Dibismuth trioxide

AC19262-0010 AC19262-0050
TS19262-0010EA 141 USD
TS19262-0010 TS19262-0050
Bismuth(III) oxide 98+%, pure
Bismuth (III) oxide
Formula: BiO₃
Boiling Pt: 1890 °C (760 mmHg)
Melting Pt: 825 °C
Density: 8.9 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now

Specification Test Results

Appearance Yellow Powder
Assay 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR