Chromium(III) oxide 99%, amorphous powder

Supplier: Thermo Scientific Chemicals

Synonyms: Chromic oxide

AC19208-0025 AC19208-5000
TS19208-0025EA 216 USD
TS19208-0025 TS19208-5000
Chromium(III) oxide 99%, amorphous powder
Chromium (III) oxide
Formula: Cr₂O₃
Boiling Pt: 4000 °C (760 mmHg)
Melting Pt: 2435 °C
Density: 5.21 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 13,02247

Order Now

Specification Test Results

Appearance Green Powder
Assay ≥98.5 %
Additional info tamped apparent density (g/ml): 1.0 to 1.3
pH 5 to 7
Loss on heating ≤0.4 % (1000°C, 0.5 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR